missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroaniline 99.0+%, TCI America™

Catalog No. C011225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C011225G 25 g
C0112500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C011225G Supplier TCI America Supplier No. C011225G

Chemical Identifiers

CAS 106-47-8
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.571
MDL Number MFCD00007835
InChI Key QSNSCYSYFYORTR-UHFFFAOYSA-N
Synonym p-chloroaniline, 4-chlorobenzenamine, p-chloraniline, benzenamine, 4-chloro, p-chlorophenylamine, 4-chlorophenylamine, para-chloroaniline, 1-amino-4-chlorobenzene, p-aminochlorobenzene, 4-aminochlorobenzene
PubChem CID 7812
ChEBI CHEBI:20331
IUPAC Name 4-chloroaniline
SMILES C1=CC(=CC=C1N)Cl

Specifications

Melting Point 72°C
Quantity 25 g
Physical Form Crystal-Powder at 20°C
UN Number 2018
Boiling Point 232°C
Formula Weight 127.57
Percent Purity ≥99.0% (GC)
Chemical Name or Material 4-Chloroaniline
EINECSNumber (3)-0194
RTECSNumber BX0700000
TSCA Yes