missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroaniline Hydrochloride 98.0+%, TCI America™

Catalog No. C0115500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0115500G 500 g
C011525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0115500G Supplier TCI America Supplier No. C0115500G

Chemical Identifiers

CAS 20265-96-7
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.029
MDL Number MFCD00042057
InChI Key ISJBQSJDQZLCSF-UHFFFAOYSA-N
Synonym 4-chloroaniline hydrochloride, p-chloroaniline hydrochloride, 4-chlorobenzenamine hydrochloride, 4-chloroanilinium chloride, benzenamine, 4-chloro-, hydrochloride, p-chloroanilinium chloride, unii-po3w01cfsw, p-chloroaniline hcl, 4-chloroaniline hcl, p-chlorophenylamine hydrochloride
PubChem CID 5284363
IUPAC Name 4-chloroaniline;hydrochloride
SMILES C1=CC(=CC=C1N)Cl.Cl

Specifications

Melting Point 240°C
Color White
Quantity 500 g
UN Number 2018
Formula Weight 164.03
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloroaniline Hydrochloride
RTECSNumber BX1394000
TSCA Yes