missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-6-hydroxypyrimidine 98.0+%, TCI America™

Catalog No. C23685G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23685G 5 g
C23681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23685G Supplier TCI America Supplier No. C23685G

Chemical Identifiers

CAS 4765-77-9
Molecular Formula C4H3ClN2O
Molecular Weight (g/mol) 130.531
MDL Number MFCD00233957
InChI Key AXFABVAPHSWFMD-UHFFFAOYSA-N
Synonym 6-chloro-4-hydroxypyrimidine, 6-chloropyrimidin-4-ol, 4-chloro-6-hydroxypyrimidine, 6-chloropyrimidin-4 1h-one, 6-chloropyrimidin-4 3h-one, 6-chloro-4-pyrimidinol, 6-chloro-4 1h-pyrimidinone, 4 1h-pyrimidinone, 6-chloro, 6-chloro-3,4-dihydro-4-oxopyrimidine, zlchem 18
PubChem CID 358318
IUPAC Name 6-chloro-1H-pyrimidin-4-one
SMILES C1=C(NC=NC1=O)Cl

Specifications

Melting Point 195°C
Color White-Yellow
Quantity 5 g
Formula Weight 130.53
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-6-hydroxypyrimidine