missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-5-fluoro-2-hydroxybenzophenone 97.0+%, TCI America™

Numéro de catalogue. C33381G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C33381G 1 g
C33385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C33381G Fournisseur TCI America Code fournisseur C33381G
Il en reste null

Identifiants chimiques

CAS 62433-26-5
Molecular Formula C13H8ClFO2
Molecular Weight (g/mol) 250.653
MDL Number MFCD00040948
InChI Key AYBQWBCUAWOLCT-UHFFFAOYSA-N
PubChem CID 112841
IUPAC Name (4-chlorophenyl)-(5-fluoro-2-hydroxyphenyl)methanone
SMILES C1=CC(=CC=C1C(=O)C2=C(C=CC(=C2)F)O)Cl

Spécifications

Melting Point 68°C
Color Yellow
Quantity 1 g
Formula Weight 250.65
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chloro-5-fluoro-2-hydroxybenzophenone