missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine, 96%

Catalog No. AAH33260MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33260MD 250 mg
AAH3326003 1 g
AAH3326006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH33260MD Supplier Thermo Scientific Chemicals Supplier No. H33260MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 331761-46-7 | C12H6Cl2N2S | 281.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 331761-46-7
Molecular Formula C12H6Cl2N2S
Molecular Weight (g/mol) 281.154
MDL Number MFCD01038375
InChI Key QDMQHIKVIOWCGY-UHFFFAOYSA-N
Synonym 4-chloro-5-4-chlorophenyl thieno 2,3-d pyrimidine, 4-chloro-5-4-chloro-phenyl-thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5-4-chlorophenyl, 4-chloro-5-4-chlorophenyl thiopheno 2,3-d pyrimidine, 4-chloranyl-5-4-chlorophenyl thieno 2,3-d pyrimidine
PubChem CID 708755
IUPAC Name 4-chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine
SMILES C1=CC(=CC=C1C2=CSC3=C2C(=NC=N3)Cl)Cl

Specifications

Melting Point 139.5°C to 141°C
Quantity 250 mg
Formula Weight 281.16
Percent Purity 96%
Chemical Name or Material 4-Chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only