missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrocoumarin 96.0+%, TCI America™

Catalog No. C16915G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16915G Supplier TCI America Supplier No. C16915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38464-20-9
Molecular Formula C9H4ClNO4
Molecular Weight (g/mol) 225.584
MDL Number MFCD00051670
InChI Key OFLRQEKOAGDHKT-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrocoumarin, 4-chloro-3-nitro-2h-chromen-2-one, 4-chloro-3-nitro-chromen-2-one, 3-nitro-4-chlorocoumarin, pubchem8672, acmc-1ahgg, 4-chloranyl-3-nitro-chromen-2-one, 4-chloro-3-nitro-1-benzopyran-2-one, 2h-1-benzopyran-2-one,4-chloro-3-nitro
PubChem CID 688979
IUPAC Name 4-chloro-3-nitrochromen-2-one
SMILES C1=CC=C2C(=C1)C(=C(C(=O)O2)[N+](=O)[O-])Cl

Specifications

Melting Point 162°C
Quantity 5 g
Formula Weight 225.58
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-nitrocoumarin