missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrocoumarin, 98%

Catalog No. AAL0790806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0790806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0790806 Supplier Thermo Scientific Chemicals Supplier No. L0790806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 38464-20-9 | Molecular Formula: C9H4ClNO4 | Molecular Weight: 225.584 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Chloro-3-nitrocoumarin, is used in synthesis and antimicrobial activity of new 4-heteroarylamino coumarin derivatives that contain nitrogen and sulfur as heteroatoms.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 38464-20-9
Molecular Formula C9H4ClNO4
Molecular Weight (g/mol) 225.584
MDL Number MFCD00051670
InChI Key OFLRQEKOAGDHKT-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrocoumarin, 4-chloro-3-nitro-2h-chromen-2-one, 4-chloro-3-nitro-chromen-2-one, 3-nitro-4-chlorocoumarin, pubchem8672, acmc-1ahgg, 4-chloranyl-3-nitro-chromen-2-one, 4-chloro-3-nitro-1-benzopyran-2-one, 2h-1-benzopyran-2-one,4-chloro-3-nitro
PubChem CID 688979
IUPAC Name 4-chloro-3-nitrochromen-2-one
SMILES C1=CC=C2C(=C1)C(=C(C(=O)O2)[N+](=O)[O-])Cl

Specifications

Melting Point ∽162°C (decomposition)
Quantity 5 g
Beilstein 883649
Solubility Information Insoluble in water.
Formula Weight 225.59
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-nitrocoumarin
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only