missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-nitrobenzonitrile 98.0+%, TCI America™
Supplier: TCI America C15485G
Specifications
| 4-Chloro-3-nitrobenzonitrile | |
| 99°C | |
| MFCD00016987 | |
| 3-nitro-4-chlorobenzonitrile, 4-chloro-3-nitro-benzonitrile, benzonitrile, 4-chloro-3-nitro, 4-chlor-3-nitrobenzonitril, 2-chloro-5-cyanonitrobenzene, 4-chlor-3-nitrobenzonitril czech, 4-chloro-3-nitro-benzonitril, chembl54162, 4-09-00-01227 beilstein handbook reference, 4-chloro-3-nitrobenzenecarbonitrile | |
| C1=CC(=C(C=C1C#N)[N+](=O)[O-])Cl | |
| 182.563 | |
| 182.56 | |
| Crystalline Powder |
| 939-80-0 | |
| C7H3ClN2O2 | |
| 5 g | |
| XBLPHYSLHRGMNW-UHFFFAOYSA-N | |
| 4-chloro-3-nitrobenzonitrile | |
| 13655 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 939-80-0 | |
| 182.563 | |
| XBLPHYSLHRGMNW-UHFFFAOYSA-N | |
| 13655 | |
| C1=CC(=C(C=C1C#N)[N+](=O)[O-])Cl |
| C7H3ClN2O2 | |
| MFCD00016987 | |
| 3-nitro-4-chlorobenzonitrile, 4-chloro-3-nitro-benzonitrile, benzonitrile, 4-chloro-3-nitro, 4-chlor-3-nitrobenzonitril, 2-chloro-5-cyanonitrobenzene, 4-chlor-3-nitrobenzonitril czech, 4-chloro-3-nitro-benzonitril, chembl54162, 4-09-00-01227 beilstein handbook reference, 4-chloro-3-nitrobenzenecarbonitrile | |
| 4-chloro-3-nitrobenzonitrile |
Safety and Handling
EINECSNumber : (3)-1781
RTECSNumber : DI3050000
TSCA : No