missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzonitrile 98.0+%, TCI America™

Catalog No. p-7125052
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C15485G 5 g
C154825G 25 g
2 options

Catalog No. C15485G

Supplier: TCI America C15485G

Only null left

Chemical Identifiers

CAS 939-80-0
Molecular Formula C7H3ClN2O2
Molecular Weight (g/mol) 182.563
MDL Number MFCD00016987
InChI Key XBLPHYSLHRGMNW-UHFFFAOYSA-N
Synonym 3-nitro-4-chlorobenzonitrile, 4-chloro-3-nitro-benzonitrile, benzonitrile, 4-chloro-3-nitro, 4-chlor-3-nitrobenzonitril, 2-chloro-5-cyanonitrobenzene, 4-chlor-3-nitrobenzonitril czech, 4-chloro-3-nitro-benzonitril, chembl54162, 4-09-00-01227 beilstein handbook reference, 4-chloro-3-nitrobenzenecarbonitrile
PubChem CID 13655
IUPAC Name 4-chloro-3-nitrobenzonitrile
SMILES C1=CC(=C(C=C1C#N)[N+](=O)[O-])Cl

Specifications

Melting Point 99°C
Quantity 5 g
Formula Weight 182.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-nitrobenzonitrile
EINECSNumber (3)-1781
RTECSNumber DI3050000
TSCA No