accessibility menu, dialog, popup

UserName

ONOClN

CAS RN 939-80-0

IUPAC Name: 4-chloro-3-nitrobenzonitrile
Synonyms: 4-chloro-3-nitrobenzonitrile
Molecular Weight (g/mol): 182.56
Molecular Formula: C7H3ClN2O2
InChi Key: XBLPHYSLHRGMNW-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1Cl)C#N
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)