missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. C0223500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0223500G 500 g
C022325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0223500G Supplier TCI America Supplier No. C0223500G

Chemical Identifiers

CAS 96-99-1
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.562
MDL Number MFCD00007079
InChI Key DFXQXFGFOLXAPO-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrobenzoic acid, 3-nitro-4-chlorobenzoic acid, benzoic acid, 4-chloro-3-nitro, kyselina 4-chloro-3-nitrobenzoova, unii-0mlz915q9k, ccris 3126, kyselina 4-chloro-3-nitrobenzoova czech, 4-chloro-3-nitrobenzoic acic, 4-chloro-3-nitro-benzoic acid, 5-carboxy-2-chloronitrobenzene
PubChem CID 7320
SMILES C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])Cl

Specifications

Melting Point 182°C
Color White-Yellow
Quantity 500 g
Formula Weight 201.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-nitrobenzoic Acid
EINECSNumber (3)-1502
RTECSNumber DG5425050
TSCA Yes