missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzenesulfonyl Chloride 96.0+%, TCI America™

Catalog No. C18875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18875G 5 g
C188725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18875G Supplier TCI America Supplier No. C18875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 97-08-5
Molecular Formula C6H3Cl2NO4S
Molecular Weight (g/mol) 256.053
InChI Key SEWNAJIUKSTYOP-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrobenzene-1-sulfonyl chloride, 4-chloro-3-nitrobenzenesulfonylchloride, benzenesulfonyl chloride, 4-chloro-3-nitro, yellow sulfon chloride, 3-nitro-4-chlorobenzenesulfonyl chloride, 4-chloro-3-nitrobenzenesulphonyl chloride, 4-chloro-3-nitro benzene sulfonyl chloride, chloro 4-chloro-3-nitrophenyl sulfone, ksc486m2p, sewnajiukstyop-uhfffaoysa
PubChem CID 7323
IUPAC Name 4-chloro-3-nitrobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)[N+](=O)[O-])Cl

Specifications

Melting Point 60°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 256.05
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-nitrobenzenesulfonyl Chloride
EINECSNumber (3)-2070
TSCA Yes