missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C30711G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C30711G Supplier TCI America Supplier No. C30711G

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material 4-Chloro-3-methylphenylboronic Acid (contains varying amounts of Anhydride)
Melting Point 258°C
CAS 161950-10-3
Color White
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Molecular Formula C7H8BClO2
MDL Number MFCD02683104
Synonym 4-chloro-3-methylbenzeneboronic acid, 4-chloro-3-methylphenyl boronic acid, 4-chloro-m-tolueneboronic acid, 4-chloro-m-tolylboronic acid, 4-chloro-3-methyl-phenyl boronic acid, boronic acid, 4-chloro-3-methylphenyl, boronic acid, b-4-chloro-3-methylphenyl, acmc-209xzw, 4-chloro-3-methylphenylboronicacid, 3-methyl-4-chlorophenylboronic acid
InChI Key UZDPQDBLCJDUAX-UHFFFAOYSA-N
SMILES CC1=C(Cl)C=CC(=C1)B(O)O
IUPAC Name (4-chloro-3-methylphenyl)boronic acid
Molecular Weight (g/mol) 170.40
PubChem CID 3786632
Formula Weight 170.40
Show More Show Less