missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3,5-dinitrobenzoic Acid 98.0+%, TCI America™
Supplier: TCI America C060725G
Specifications
| 4-Chloro-3,5-dinitrobenzoic Acid | |
| 160°C | |
| C7H3ClN2O6 | |
| 25 g | |
| PCTFIHOVQYYAMH-UHFFFAOYSA-N | |
| 246.559 | |
| 246.56 | |
| Crystalline Powder |
| 118-97-8 | |
| Yellow | |
| MFCD00007080 | |
| 4-chloro-3,5-dinitrobenzoic acid, 3,5-dinitro-4-chlorobenzoic acid, benzoic acid, 4-chloro-3,5-dinitro, 4-chloro-3,5-dinitrobenzoate, 4-chloro-3,5-dinitro-benzoic acid, benzoicacid, 4-chloro-3,5-dinitro, pubchem22980, acmc-1bt9j, ksc180k5t, timtec-bb sbb003176 | |
| C1=C(C=C(C(=C1[N+](=O)[O-])Cl)[N+](=O)[O-])C(=O)O | |
| 8377 | |
| ≥98.0% (T) |
Chemical Identifiers
| 118-97-8 | |
| 246.559 | |
| PCTFIHOVQYYAMH-UHFFFAOYSA-N | |
| 8377 |
| C7H3ClN2O6 | |
| MFCD00007080 | |
| 4-chloro-3,5-dinitrobenzoic acid, 3,5-dinitro-4-chlorobenzoic acid, benzoic acid, 4-chloro-3,5-dinitro, 4-chloro-3,5-dinitrobenzoate, 4-chloro-3,5-dinitro-benzoic acid, benzoicacid, 4-chloro-3,5-dinitro, pubchem22980, acmc-1bt9j, ksc180k5t, timtec-bb sbb003176 | |
| C1=C(C=C(C(=C1[N+](=O)[O-])Cl)[N+](=O)[O-])C(=O)O |
Safety and Handling
TSCA : Yes