missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3,5-dinitrobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7124522
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C060725G 25 g
C0607250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C060725G Supplier TCI America Supplier No. C060725G
Only null left

Chemical Identifiers

CAS 118-97-8
Molecular Formula C7H3ClN2O6
Molecular Weight (g/mol) 246.559
MDL Number MFCD00007080
InChI Key PCTFIHOVQYYAMH-UHFFFAOYSA-N
Synonym 4-chloro-3,5-dinitrobenzoic acid, 3,5-dinitro-4-chlorobenzoic acid, benzoic acid, 4-chloro-3,5-dinitro, 4-chloro-3,5-dinitrobenzoate, 4-chloro-3,5-dinitro-benzoic acid, benzoicacid, 4-chloro-3,5-dinitro, pubchem22980, acmc-1bt9j, ksc180k5t, timtec-bb sbb003176
PubChem CID 8377
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])Cl)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 160°C
Color Yellow
Quantity 25 g
Formula Weight 246.56
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3,5-dinitrobenzoic Acid