accessibility menu, dialog, popup

UserName

HOOClNOONOO

CAS RN 118-97-8

IUPAC Name: 4-chloro-3,5-dinitrobenzoic acid
Synonyms: 4-chloro-3,5-dinitrobenzoic acid
Molecular Weight (g/mol): 246.56
Molecular Formula: C7H3ClN2O6
InChi Key: PCTFIHOVQYYAMH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=C(Cl)C(=C1)[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)