missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-phenylquinoline, 99%

Catalog No. AC164870025
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164870025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC164870025 Supplier Thermo Scientific Chemicals Supplier No. 164870025
Only null left

CAS: 4979-79-7 | C15H10ClN | 239.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4979-79-7
Molecular Formula C15H10ClN
Molecular Weight (g/mol) 239.71
MDL Number MFCD00160641
InChI Key GLVDSTVYOFXBKT-UHFFFAOYSA-N
Synonym 4-chloro-2-phenyl-quinoline, 4-chloro-2-phenyquinoline, 2-phenyl-4-chloroquinoline, 4-chloro-2-phenyl-quinolin, 4-chloro-2-phenyl quinoline
PubChem CID 278376
IUPAC Name 4-chloro-2-phenylquinoline
SMILES C1=CC=C(C=C1)C2=NC3=CC=CC=C3C(=C2)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 62°C to 64°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 2.5 g
Formula Weight 239.71
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Chloro-2-phenylquinoline, 99%
Show More Show Less

RUO – Research Use Only