missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-nitrotoluene, TCI America™

Catalog No. C023125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C023125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C023125G Supplier TCI America Supplier No. C023125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89-59-8
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007215
InChI Key SQFLFRQWPBEDHM-UHFFFAOYSA-N
Synonym 4-chloro-2-nitrotoluene, 2-nitro-4-chlorotoluene, benzene, 4-chloro-1-methyl-2-nitro, 4,2-chloronitrotoluene, p-chloro-o-nitrotoluene, toluene, 4-chloro-2-nitro, 4-chloro-1-methyl-2-nitro-benzene, ccris 3116, 4-chlor-2-nitrotoluol, pubchem18919
PubChem CID 6975
IUPAC Name 4-chloro-1-methyl-2-nitrobenzene
SMILES CC1=CC=C(Cl)C=C1[N+]([O-])=O

Specifications

Melting Point 38°C
Color Yellow
Boiling Point 115°C
Quantity 25 g
UN Number 3457
Formula Weight 171.58
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-nitrotoluene
EINECSNumber (3)-0449
TSCA Yes