missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylquinoline 98.0+%, TCI America™

Catalog No. C29015G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C29015G Supplier TCI America Supplier No. C29015G

Chemical Identifiers

CAS 4295-06-1
Molecular Formula C10H8ClN
Molecular Weight (g/mol) 177.63
MDL Number MFCD00006757
InChI Key HQAIROMRVBVWSK-UHFFFAOYSA-N
Synonym 4-chloroquinaldine, quinoline, 4-chloro-2-methyl, 4-chloro-2-methyl-quinoline, 2-methyl-4-chloroquinoline, quinaldine, 4-chloro, 4-chloro-quinaldine, acmc-20a2ui, 4-chloroquinaldine hcl, quinoline,4-chloro-2-methyl
PubChem CID 77973
IUPAC Name 4-chloro-2-methylquinoline
SMILES CC1=NC2=CC=CC=C2C(=C1)Cl

Specifications

Color Yellow
Boiling Point 270°C
Quantity 5 g
Formula Weight 177.63
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-methylquinoline