missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Numéro de catalogue. p-7030461
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAH2887209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AAH2887209 Fournisseur Thermo Scientific Chemicals Code fournisseur H2887209
Il en reste null

Used as intermediates for pharmaceutical and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as intermediates for pharmaceutical and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Identifiants chimiques

CAS 160591-91-3
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.362
MDL Number MFCD02684293
InChI Key YBNDRTRLXPEWKQ-UHFFFAOYSA-N
Synonym 4-chloro-2-fluorobenzeneboronic acid, 4-chloro-2-fluorophenyl boronic acid, 4-chloro-2-fluorophenylboronicacid, 2-fluoro-4-chlorophenylboronic acid, b-4-chloro-2-fluorophenyl-boronic acid, 4-chloro-2-fluorophenyl-boronic acid, boronic acid, 4-chloro-2-fluorophenyl, boronic acid, b-4-chloro-2-fluorophenyl, pubchem5034, 4-chloro-2-fluoro-phenyl boronic acid
PubChem CID 4193595
IUPAC Name (4-chloro-2-fluorophenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)Cl)F)(O)O

Spécifications

Melting Point 246°C to 250°C
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 174.37
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-fluorobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only