missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,6-dimethyl-3-nitropyridine 98.0+%, TCI America™

Catalog No. C31461G
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
C31461G 1 g
1 options

Catalog No. C31461G

Supplier: TCI America C31461G

Only null left

Chemical Identifiers

CAS 15513-48-1
Molecular Formula C7H7ClN2O2
Molecular Weight (g/mol) 186.595
MDL Number MFCD08543468
InChI Key PSBGFLWVQDGETK-UHFFFAOYSA-N
Synonym pyridine, 4-chloro-2,6-dimethyl-3-nitro, 4-chloro-3-nitro-2,6-lutidine, pubchem11177, 4-chloranyl-2,6-dimethyl-3-nitro-pyridine
PubChem CID 10702725
IUPAC Name 4-chloro-2,6-dimethyl-3-nitropyridine
SMILES CC1=NC(=C(C(=C1)Cl)[N+](=O)[O-])C

Specifications

Melting Point 73°C
Color White-Yellow
Quantity 1 g
Formula Weight 186.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2,6-dimethyl-3-nitropyridine