missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,5-dimethoxynitrobenzene 98.0+%, TCI America™

Catalog No. C124425G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C124425G 25 g
C1244250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C124425G Supplier TCI America Supplier No. C124425G

Chemical Identifiers

CAS 6940-53-0
Molecular Formula C8H8ClNO4
Molecular Weight (g/mol) 217.605
MDL Number MFCD00024577
InChI Key ORLPGMKKCAEWOW-UHFFFAOYSA-N
PubChem CID 81362
IUPAC Name 1-chloro-2,5-dimethoxy-4-nitrobenzene
SMILES COC1=CC(=C(C=C1[N+](=O)[O-])OC)Cl

Specifications

Melting Point 141°C
Quantity 25 g
Formula Weight 217.61
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2,5-dimethoxynitrobenzene