missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-chloro-2,5-dimethoxyaniline, 97%, Thermo Scientific™
Specifications
Specifications
| Nom ou substance chimique | 4-chloro-2, 5-dimethoxyaniline |
| Point d’éclair | 163°C |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 96% min. (HPLC) |
| Emballage | Glass bottle |
| Quantité | 100g |
| Poids de formule | 187.62 |
| Pourcentage de pureté | 97% |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. |
| EINECSNumber | 228-782- |
RUO – Research Use Only