missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-2,2,2-trifluoroacetophenone 97.0+%, TCI America™

Numéro de catalogue. C17381G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C17381G 1 g
C17385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C17381G Fournisseur TCI America Code fournisseur C17381G
Il en reste null

Identifiants chimiques

CAS 321-37-9
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.564
MDL Number MFCD00018686
InChI Key DYPQUENOGZXOGE-UHFFFAOYSA-N
PubChem CID 67578
IUPAC Name 1-(4-chlorophenyl)-2,2,2-trifluoroethanone
SMILES C1=CC(=CC=C1C(=O)C(F)(F)F)Cl

Spécifications

Color White-Yellow
Quantity 1 g
Formula Weight 208.56
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4′-Chloro-2,2,2-trifluoroacetophenone