missing translation for 'onlineSavingsMsg'
Learn More

4-Carbamoylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C25701G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C25701G Supplier TCI America Supplier No. C25701G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-Carbamoylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 123088-59-5
Melting Point 234°C
Molecular Formula C7H8BNO3
Quantity 1 g
MDL Number MFCD03411940
Synonym 4-aminocarbonylphenylboronic acid, 4-carbamoylphenyl boronic acid, benzamide-4-boronic acid, 4-aminocarbonyl benzeneboronic acid, 4-carbamoylbenzeneboronic acid, 4-aminocarbonylphenyl boronic acid, 4-dihydroxyboranyl benzamide, p-aminocarbonyl phenylboronic acid, boronic acid, 4-aminocarbonyl phenyl, 4-aminocarbonyl phenylboronic acid
InChI Key GNRHNKBJNUVWFZ-UHFFFAOYSA-N
SMILES NC(=O)C1=CC=C(C=C1)B(O)O
IUPAC Name (4-carbamoylphenyl)boronic acid
Molecular Weight (g/mol) 164.96
PubChem CID 2737811
Formula Weight 164.96
Physical Form Crystalline Powder
Show More Show Less