missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Butyrylbiphenyl 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 95°C |
| Boiling Point | 355°C |
| Quantity | 500 g |
| Synonym | 4′-Phenylbutyrophenone |
| Formula Weight | 224.30 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Butyrylbiphenyl |
Safety and Handling
| TSCA | No |