missing translation for 'onlineSavingsMsg'
Learn More

4-Butylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B25891G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B25891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B25891G Supplier TCI America Supplier No. B25891G
Only null left

Chemical Identifiers

CAS 145240-28-4
Molecular Formula C10H15BO2
Molecular Weight (g/mol) 178.038
MDL Number MFCD02093926
InChI Key UGZUUTHZEATQAM-UHFFFAOYSA-N
PubChem CID 4100860
IUPAC Name (4-butylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)CCCC)(O)O

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 1 g
Synonym 4-butylphenyl boronic acid, 4-n-butylphenylboronic acid, 4-butylphenylboronicacid, 4-butylbenzeneboronic acid, 4-n-butylbenzeneboronic acid, 4-butylphenyl boranediol, boronic acid, 4-butylphenyl, pubchem18523, acmc-209cue, 4-butylphenylboranic acid
Formula Weight 178.04
Physical Form Crystalline Powder
Chemical Name or Material 4-Butylphenylboronic Acid (contains varying amounts of Anhydride)