missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Butylbenzaldehyde 95.0+%, TCI America™
Supplier: TCI America B101425ML
Specifications
| 4-Butylbenzaldehyde | |
| Yellow | |
| C11H14O | |
| 25 mL | |
| ARIREUPIXAKDAY-UHFFFAOYSA-N | |
| 4-butylbenzaldehyde | |
| 70988 | |
| ≥95.0% (GC) |
| 1200-14-2 | |
| 165°C | |
| MFCD00040751 | |
| benzaldehyde, 4-butyl, p-butylbenzaldehyde, 4-n-butylbenzaldehyde, butylbenzaldehyde, p-n-butylbenzaldehyde, butylbenzaldehyde;, 4-butylbenzaldehyd, 4-butyl-benzaldehyde, p-butyl-benzaldehyde, h | |
| CCCCC1=CC=C(C=O)C=C1 | |
| 162.23 | |
| 162.23 | |
| Liquid |
Chemical Identifiers
| 1200-14-2 | |
| 162.23 | |
| ARIREUPIXAKDAY-UHFFFAOYSA-N | |
| 70988 | |
| CCCCC1=CC=C(C=O)C=C1 |
| C11H14O | |
| MFCD00040751 | |
| benzaldehyde, 4-butyl, p-butylbenzaldehyde, 4-n-butylbenzaldehyde, butylbenzaldehyde, p-n-butylbenzaldehyde, butylbenzaldehyde;, 4-butylbenzaldehyd, 4-butyl-benzaldehyde, p-butyl-benzaldehyde, h | |
| 4-butylbenzaldehyde |
Safety and Handling
TSCA : Yes