accessibility menu, dialog, popup

UserName

H3CO

CAS RN 1200-14-2

IUPAC Name: 4-butylbenzaldehyde
Synonyms: 4-butylbenzaldehyde
Molecular Weight (g/mol): 162.23
Molecular Formula: C11H14O
InChi Key: ARIREUPIXAKDAY-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=O)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results