missing translation for 'onlineSavingsMsg'
Learn More

4-Butylbenzaldehyde 95.0+%, TCI America™

Catalog No. p-7118804
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10145ML 5 mL
B101425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B10145ML Supplier TCI America Supplier No. B10145ML
Only null left

Chemical Identifiers

CAS 1200-14-2
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00040751
InChI Key ARIREUPIXAKDAY-UHFFFAOYSA-N
Synonym benzaldehyde, 4-butyl, p-butylbenzaldehyde, 4-n-butylbenzaldehyde, butylbenzaldehyde, p-n-butylbenzaldehyde, butylbenzaldehyde;, 4-butylbenzaldehyd, 4-butyl-benzaldehyde, p-butyl-benzaldehyde, h
PubChem CID 70988
IUPAC Name 4-butylbenzaldehyde
SMILES CCCCC1=CC=C(C=O)C=C1

Specifications

Color Yellow
Boiling Point 165°C
Quantity 5 mL
Formula Weight 162.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Butylbenzaldehyde