missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Butyl-4-iodobiphenyl 98.0+%, TCI America™
$137.94 - $408.22
Chemical Identifiers
| CAS | 199982-02-0 |
|---|---|
| Molecular Formula | C16H17I |
| Molecular Weight (g/mol) | 336.216 |
| MDL Number | MFCD16619281 |
| InChI Key | KTEKHPDWAMYUBP-UHFFFAOYSA-N |
| PubChem CID | 53417225 |
| IUPAC Name | 1-butyl-4-(4-iodophenyl)benzene |
| SMILES | CCCCC1=CC=C(C=C1)C2=CC=C(C=C2)I |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B5211200MG
|
TCI America
B5211200MG |
200 mg |
Each for $137.94
|
|
|||||
|
B52111G
|
TCI America
B52111G |
1 g |
Each for $408.22
|
|
|||||
Chemical Identifiers
| 199982-02-0 | |
| 336.216 | |
| KTEKHPDWAMYUBP-UHFFFAOYSA-N | |
| 1-butyl-4-(4-iodophenyl)benzene |
| C16H17I | |
| MFCD16619281 | |
| 53417225 | |
| CCCCC1=CC=C(C=C1)C2=CC=C(C=C2)I |
Specifications
| 199982-02-0 | |
| White-Yellow | |
| MFCD16619281 | |
| KTEKHPDWAMYUBP-UHFFFAOYSA-N | |
| 1-butyl-4-(4-iodophenyl)benzene | |
| 53417225 | |
| ≥98.0% (GC) | |
| 4′-Butyl-4-iodobiphenyl |
| 120°C | |
| C16H17I | |
| 200 mg | |
| CCCCC1=CC=C(C=C1)C2=CC=C(C=C2)I | |
| 336.216 | |
| 336.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : No