accessibility menu, dialog, popup

UserName

CAS RN 199982-02-0

IUPAC Name: 4-butyl-4'-iodo-1,1'-biphenyl
Synonyms: 4-butyl-4'-iodo-1,1'-biphenyl
Molecular Weight (g/mol): 336.22
Molecular Formula: C16H17I
InChi Key: KTEKHPDWAMYUBP-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)C1=CC=C(I)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results