missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Butyl-4-iodobiphenyl 98.0+%, TCI America™
Supplier: TCI America B52111G
Specifications
| 4′-Butyl-4-iodobiphenyl | |
| 120°C | |
| C16H17I | |
| 1 g | |
| CCCCC1=CC=C(C=C1)C2=CC=C(C=C2)I | |
| 336.216 | |
| 336.22 | |
| Crystalline Powder |
| 199982-02-0 | |
| White-Yellow | |
| MFCD16619281 | |
| KTEKHPDWAMYUBP-UHFFFAOYSA-N | |
| 1-butyl-4-(4-iodophenyl)benzene | |
| 53417225 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 199982-02-0 | |
| 336.216 | |
| KTEKHPDWAMYUBP-UHFFFAOYSA-N | |
| 1-butyl-4-(4-iodophenyl)benzene |
| C16H17I | |
| MFCD16619281 | |
| 53417225 | |
| CCCCC1=CC=C(C=C1)C2=CC=C(C=C2)I |
Safety and Handling
TSCA : No