missing translation for 'onlineSavingsMsg'
Learn More

4-Butoxyphthalonitrile 96.0+%, TCI America™

Catalog No. B13531G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B13531G Supplier TCI America Supplier No. B13531G
Only null left

Specifications

CAS 81560-32-9
Melting Point 46°C
Color Yellow
Molecular Formula C12H12N2O
Quantity 1 g
MDL Number MFCD00059035
Synonym 4-Butoxy-1,2-dicyanobenzene
InChI Key AVGCGXBXMPWLOW-UHFFFAOYSA-N
SMILES CCCCOC1=CC(=C(C=C1)C#N)C#N
IUPAC Name 4-butoxybenzene-1,2-dicarbonitrile
Molecular Weight (g/mol) 200.241
PubChem CID 546656
Formula Weight 200.24
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Butoxyphthalonitrile
Show More Show Less