missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoveratrole, 97%

Catalog No. AAA1113336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1113336 500 g
AAA1113314 25 g
AAA1113322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1113336 Supplier Thermo Scientific Chemicals Supplier No. A1113336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2859-78-1 | C8H9BrO2 | 217.062 g/mol

4-Bromo-1,2-dimethoxybenzene is a metabolite of bromobenzene, with a catechol moiety in the substructure. It also has use as a redox shuttle additive, essentially a component in lithium batteries that consumes excess current during overcharge. And also used in the synthesis of the isoquinoline alkaloid (-)-mesembrine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-1,2-dimethoxybenzene is a metabolite of bromobenzene, with a catechol moiety in the substructure. It also has use as a redox shuttle additive, essentially a component in lithium batteries that consumes excess current during overcharge. And also used in the synthesis of the isoquinoline alkaloid (-)-mesembrine.

Solubility
Soluble in benzene, ether, toluene.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 2859-78-1
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.062
MDL Number MFCD00008381
InChI Key KBTMGSMZIKLAHN-UHFFFAOYSA-N
Synonym 4-bromoveratrole, 1-bromo-3,4-dimethoxybenzene, 3,4-dimethoxybromobenzene, p-bromoveratrole, benzene, 4-bromo-1,2-dimethoxy, 3,4-dimethoxyphenyl bromide, 4-bromo-1,2-dimethoxy-benzene, 1,2-dimethoxy-4-bromobenzene, 4-bromocatechol dimethyl ether, 4-bromoveratrol
PubChem CID 76114
IUPAC Name 4-bromo-1,2-dimethoxybenzene
SMILES COC1=C(C=C(C=C1)Br)OC

Specifications

Density 1.51
Boiling Point 255°C to 256°C
Flash Point 109°C (228°F)
Refractive Index 1.573
Quantity 500 g
Beilstein 2046884
Solubility Information Soluble in benzene,ether,toluene.
Formula Weight 217.06
Percent Purity 97%
Chemical Name or Material 4-Bromoveratrole
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 220-677-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only