missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromoveratrole, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 107541000
| Quantity | 100g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C8H9BrO2 | |
| MFCD00008381 | |
| 4-bromoveratrole, 1-bromo-3,4-dimethoxybenzene, 3,4-dimethoxybromobenzene, p-bromoveratrole, benzene, 4-bromo-1,2-dimethoxy, 3,4-dimethoxyphenyl bromide, 4-bromo-1,2-dimethoxy-benzene, 1,2-dimethoxy-4-bromobenzene, 4-bromocatechol dimethyl ether, 4-bromoveratrol | |
| 4-bromo-1,2-dimethoxybenzene |
Specifications
| 4-Bromoveratrole | |
| 2859-78-1 | |
| 100.0 | |
| 255°C to 256°C | |
| Authentic | |
| Glass bottle | |
| 1.5720 to 1.574 | |
| 100g | |
| 1.5 | |
| KBTMGSMZIKLAHN-UHFFFAOYSA-N | |
| 4-bromo-1,2-dimethoxybenzene | |
| 76114 | |
| 97% |
| 97% | |
| 96.0 | |
| 1.5000g/mL | |
| 109°C | |
| 96% min. (GC) | |
| C8H9BrO2 | |
| BrC6H3(OCH3)2 | |
| MFCD00008381 | |
| 4-bromoveratrole, 1-bromo-3,4-dimethoxybenzene, 3,4-dimethoxybromobenzene, p-bromoveratrole, benzene, 4-bromo-1,2-dimethoxy, 3,4-dimethoxyphenyl bromide, 4-bromo-1,2-dimethoxy-benzene, 1,2-dimethoxy-4-bromobenzene, 4-bromocatechol dimethyl ether, 4-bromoveratrol | |
| COC1=C(C=C(C=C1)Br)OC | |
| 217.06 | |
| 217.06 |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 220-677-8
TSCA : TSCA
Promotions
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.