missing translation for 'onlineSavingsMsg'
Learn More

4-Bromothioanisole, 98+%

Catalog No. p-7022634
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1550414 25 g
AAA1550422 100 g
AAA1550436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1550414 Supplier Thermo Scientific Chemicals Supplier No. A1550414

CAS: 104-95-0 | C7H7BrS | 203.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-95-0
Molecular Formula C7H7BrS
Molecular Weight (g/mol) 203.10
MDL Number MFCD00000102
InChI Key YEUYZNNBXLMFCW-UHFFFAOYSA-N
Synonym 4-bromothioanisole, 1-bromo-4-methylthio benzene, p-bromophenyl methyl sulfide, 4-bromophenyl methyl sulfide, benzene, 1-bromo-4-methylthio, 1-bromo-4-methylsulfanyl benzene, p-bromothioanisole, 4-bromo-thioanisole, methyl 4-bromophenylsulfide, p-bromo thioanisole
PubChem CID 66037
IUPAC Name 1-bromo-4-methylsulfanylbenzene
SMILES CSC1=CC=C(Br)C=C1

Specifications

Melting Point 37°C to 40°C
Boiling Point 250°C
Flash Point >110°C (230°F)
Odor Stench
Quantity 25 g
Beilstein 1906693
Formula Weight 203.11
Percent Purity ≥98%
Chemical Name or Material 4-Bromothioanisole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 6; Packaging Group: III
EINECSNumber 203-255-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only