missing translation for 'onlineSavingsMsg'
Learn More

4-Bromostyrene (stabilized with TBC) 95.0+%, TCI America™

Catalog No. B19805G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19805G 5 g
B198025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19805G Supplier TCI America Supplier No. B19805G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2039-82-9
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.05
MDL Number MFCD00000110
InChI Key WGGLDBIZIQMEGH-UHFFFAOYSA-N
Synonym 4-bromostyrene, 1-bromo-4-vinylbenzene, p-bromostyrene, benzene, 1-bromo-4-ethenyl, styrene, p-bromo, 1-4-bromophenyl ethylene, unii-tvk4s80xlu, p-bromo styrene, 1-bromo-4-vinyl-benzene, tvk4s80xlu
PubChem CID 16263
IUPAC Name 1-bromo-4-ethenylbenzene
SMILES BrC1=CC=C(C=C)C=C1

Specifications

Color Yellow
Boiling Point 81°C
Quantity 5 g
Formula Weight 183.05
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromostyrene (stabilized with TBC)
TSCA No
Recommended Storage Freezer