missing translation for 'onlineSavingsMsg'
Learn More

4-Bromopyridine Hydrochloride 98.0+%, TCI America™

Catalog No. B06535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B06535G 5 g
B065325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B06535G Supplier TCI America Supplier No. B06535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19524-06-2
Molecular Formula C5H5BrClN
Molecular Weight (g/mol) 194.456
MDL Number MFCD00012828
InChI Key MPZMVUQGXAOJIK-UHFFFAOYSA-N
Synonym 4-bromopyridine hydrochloride, 4-bromopyridinium chloride, 4-bromopyridine hcl, pyridine, 4-bromo-, hydrochloride, 4-bromo-pyridine hcl, 4-bromo pyridine hcl, 4-bromopyridine, chloride, 4-bromopyridinehydrochloride, 4-bromopyridine hydrochloride salt, 4-bromopyridine-hcl
PubChem CID 88100
IUPAC Name 4-bromopyridine;hydrochloride
SMILES C1=CN=CC=C1Br.Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 194.46
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromopyridine Hydrochloride