missing translation for 'onlineSavingsMsg'
Learn More

4-Bromopyridine Hydrochloride, 98%

Catalog No. AC107420250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107420250 25 g Glass bottle
AC107420050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC107420250 Supplier Thermo Scientific Chemicals Supplier No. 107420250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19524-06-2 | C5H4BrN·HCl | 194.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19524-06-2
Molecular Formula C5H4BrN·HCl
Molecular Weight (g/mol) 194.46
MDL Number MFCD00012828
InChI Key MPZMVUQGXAOJIK-UHFFFAOYSA-N
Synonym 4-bromopyridine hydrochloride, 4-bromopyridinium chloride, 4-bromopyridine hcl, pyridine, 4-bromo-, hydrochloride, 4-bromo-pyridine hcl, 4-bromo pyridine hcl, 4-bromopyridine, chloride, 4-bromopyridinehydrochloride, 4-bromopyridine hydrochloride salt, 4-bromopyridine-hcl
PubChem CID 88100
IUPAC Name 4-bromopyridine;hydrochloride
SMILES C1=CN=CC=C1Br.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 313.0°C to 315.0°C
Color Peach to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C5H4N)Br·HCl
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 194.46
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Bromopyridine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physicia
EINECSNumber 243-128-4

RUO – Research Use Only