missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophthalimide, 97+%

Catalog No. AAH6413614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6413614 25 g
AAH6413606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6413614 Supplier Thermo Scientific Chemicals Supplier No. H6413614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6941-75-9 | C8H4BrNO2 | 226.029 g/mol

It is used as a intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a intermediate for pharmaceutical and organic synthesis.

Solubility
Soluble in N,N-DMF almost transparency.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 6941-75-9
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.029
MDL Number MFCD00466049
InChI Key GNYICZVGHULCHE-UHFFFAOYSA-N
Synonym 4-bromophthalimide, 5-bromoisoindoline-1,3-dione, 5-bromo-isoindole-1,3-dione, 5-bromo-2,3-dihydro-1h-isoindole-1,3-dione, 4-bromo phthalimide, 5-bromo-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 5-bromo, 5-bromo-2h-isoindole-1,3-dione, 5-bromophthalimide, acmc-209o8b
PubChem CID 236018
IUPAC Name 5-bromoisoindole-1,3-dione
SMILES C1=CC2=C(C=C1Br)C(=O)NC2=O

Specifications

Melting Point 230°C to 235°C
Color White
Quantity 25 g
Beilstein 131235
Solubility Information Soluble in N,N-DMF almost transparency.
Formula Weight 226.03
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 4-Bromophthalimide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only