missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylacetylene, 97%

Catalog No. AAL19082MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19082MD 250 mg
AAL1908203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19082MD Supplier Thermo Scientific Chemicals Supplier No. L19082MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 766-96-1 | C8H5Br | 181.032 g/mol

4-Bromophenylacetylene, is used as the starting material for second-order nonlinear optical materials, heterocyclotriynes, and unsymmetrical 1,4-diarylbutadiynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromophenylacetylene, is used as the starting material for second-order nonlinear optical materials, heterocyclotriynes, and unsymmetrical 1,4-diarylbutadiynes.

Solubility
Soluble in chloroform, diethyl ether and ethanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 766-96-1
Molecular Formula C8H5Br
Molecular Weight (g/mol) 181.032
MDL Number MFCD00168822
InChI Key LTLVZQZDXQWLHU-UHFFFAOYSA-N
Synonym 4-bromophenylacetylene, benzene, 1-bromo-4-ethynyl, 1-bromo-4-ethynyl-benzene, 4-bromophenyl acetylene, 4'-bromophenyl acetylene, 4'-bromophenylacetylene, p-bromophenylacetylene, p-bromophenyl acetylene, 4-bromphenyl acetylene, 4-bromo-1-ethynylbenzene
PubChem CID 136603
IUPAC Name 1-bromo-4-ethynylbenzene
SMILES C#CC1=CC=C(C=C1)Br

Specifications

Melting Point 64°C to 67°C
Boiling Point 88°C to 90°C (16 mmHg)
Odor Odorless
Quantity 250 mg
Beilstein 1209726
Solubility Information Soluble in chloroform,diethyl ether and ethanol.
Formula Weight 181.04
Percent Purity 97%
Chemical Name or Material 4-Bromophenylacetylene
Hazard Category H302-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only