Learn More
4-Bromophenylacetone, 98%
CAS: 6186-22-7 | C24H24N2O5S | 452.53 g/mol
Supplier: Thermo Scientific Chemicals A1902806
Description
4-Bromophenylacetone is used to prepare biphenyl-4-yl-acetone by reacting with phenylboronic acid using cesium fluoride as a reagent and palladium phosphine complex as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromophenylacetone is used to prepare biphenyl-4-yl-acetone by reacting with phenylboronic acid using cesium fluoride as a reagent and palladium phosphine complex as a catalyst.
Solubility
Miscible with chloroform and dichloromethane. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents, reducing agents and bases.
Specifications
| 4-Bromophenylacetone | |
| C24H24N2O5S | |
| 5 g | |
| Miscible with chloroform and dichloromethane. Slightly miscible with water. | |
| COC(=O)C1=CC=CC=C1NC(=O)CN(C1=CC(C)=CC(C)=C1)S(=O)(=O)C1=CC=CC=C1 | |
| 452.53 | |
| 213.07 |
| 6186-22-7 | |
| MFCD00210401 | |
| 4-bromophenylacetone, 1-4-bromophenyl propan-2-one, 1-4-bromophenyl acetone, 1-4-bromophenyl-2-propanone, 1-4-bromo-phenyl-propan-2-one, p-bromo benzyl methyl ketone, 4-bromophenylaceton, 4-bromophenylacteone, 4-bromophenyl acetone | |
| CDUPPXGKUKTGLU-UHFFFAOYSA-N | |
| 1-(4-bromophenyl)propan-2-one | |
| 736332 | |
| 98% |
Chemical Identifiers
| 6186-22-7 | |
| 452.53 | |
| CDUPPXGKUKTGLU-UHFFFAOYSA-N | |
| 736332 | |
| COC(=O)C1=CC=CC=C1NC(=O)CN(C1=CC(C)=CC(C)=C1)S(=O)(=O)C1=CC=CC=C1 |
| C24H24N2O5S | |
| MFCD00210401 | |
| 4-bromophenylacetone, 1-4-bromophenyl propan-2-one, 1-4-bromophenyl acetone, 1-4-bromophenyl-2-propanone, 1-4-bromo-phenyl-propan-2-one, p-bromo benzyl methyl ketone, 4-bromophenylaceton, 4-bromophenylacteone, 4-bromophenyl acetone | |
| 1-(4-bromophenyl)propan-2-one |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only