missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylacetone, 98%

Catalog No. p-7024413
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1902806 5 g
AAA1902814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1902806 Supplier Thermo Scientific Chemicals Supplier No. A1902806
Only null left

CAS: 6186-22-7 | C24H24N2O5S | 452.53 g/mol

4-Bromophenylacetone is used to prepare biphenyl-4-yl-acetone by reacting with phenylboronic acid using cesium fluoride as a reagent and palladium phosphine complex as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromophenylacetone is used to prepare biphenyl-4-yl-acetone by reacting with phenylboronic acid using cesium fluoride as a reagent and palladium phosphine complex as a catalyst.

Solubility
Miscible with chloroform and dichloromethane. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, reducing agents and bases.

Chemical Identifiers

CAS 6186-22-7
Molecular Formula C24H24N2O5S
Molecular Weight (g/mol) 452.53
MDL Number MFCD00210401
InChI Key CDUPPXGKUKTGLU-UHFFFAOYSA-N
Synonym 4-bromophenylacetone, 1-4-bromophenyl propan-2-one, 1-4-bromophenyl acetone, 1-4-bromophenyl-2-propanone, 1-4-bromo-phenyl-propan-2-one, p-bromo benzyl methyl ketone, 4-bromophenylaceton, 4-bromophenylacteone, 4-bromophenyl acetone
PubChem CID 736332
IUPAC Name 1-(4-bromophenyl)propan-2-one
SMILES COC(=O)C1=CC=CC=C1NC(=O)CN(C1=CC(C)=CC(C)=C1)S(=O)(=O)C1=CC=CC=C1

Specifications

Quantity 5 g
Solubility Information Miscible with chloroform and dichloromethane. Slightly miscible with water.
Formula Weight 213.07
Percent Purity 98%
Chemical Name or Material 4-Bromophenylacetone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only