accessibility menu, dialog, popup

UserName

CH3OONHONH3CCH3SOO

CAS RN 6186-22-7

IUPAC Name: methyl 2-{2-[N-(3,5-dimethylphenyl)benzenesulfonamido]acetamido}benzoate
Synonyms: methyl 2-{2-[N-(3,5-dimethylphenyl)benzenesulfonamido]acetamido}benzoate
Molecular Weight (g/mol): 452.53
Molecular Formula: C24H24N2O5S
InChi Key: CDUPPXGKUKTGLU-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1NC(=O)CN(C1=CC(C)=CC(C)=C1)S(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)