missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenyl Isothiocyanate 98.0+%, TCI America™

Catalog No. I05265G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05265G 5 g
I052625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05265G Supplier TCI America Supplier No. I05265G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1985-12-2
Molecular Formula C7H4BrNS
Molecular Weight (g/mol) 214.08
MDL Number MFCD00004808
InChI Key XQACWEBGSZBLRG-UHFFFAOYSA-N
Synonym 4-bromophenyl isothiocyanate, trichofytocid, p-bromophenyl isothiocyanate, 4-bromophenylisothiocyanate, benzene, 1-bromo-4-isothiocyanato, bromobenzene-4-isothiocyanate, isothiocyanic acid, p-bromophenyl ester, acmc-1bqku, wln: scnr de, p-bromophenylisothiocyanate
PubChem CID 16133
IUPAC Name 1-bromo-4-isothiocyanatobenzene
SMILES C1=CC(=CC=C1N=C=S)Br

Specifications

Melting Point 58°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 214.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromophenyl Isothiocyanate
RTECSNumber NX8400000
TSCA No