missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenyl Cyclopropyl Ketone 95.0+%, TCI America™

Numéro de catalogue. B40765G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B40765G 5 g
B40761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B40765G Fournisseur TCI America Code fournisseur B40765G
Il en reste null

Identifiants chimiques

CAS 6952-89-2
Molecular Formula C10H9BrO
Molecular Weight (g/mol) 225.085
MDL Number MFCD00019227
InChI Key QTHHOINSCNBYQO-UHFFFAOYSA-N
Synonym 4-bromophenyl cyclopropyl methanone, 4-bromophenyl cyclopropyl ketone, 4-bromophenyl cyclopropylmethanone, methanone, 4-bromophenyl cyclopropyl, 4-bromo-cyclopropylcarbonylbenzene, acmc-1bag8, 4-bromobenzoyl cyclopropane, bromophenylcyclopropylmethanone
PubChem CID 81394
IUPAC Name (4-bromophenyl)-cyclopropylmethanone
SMILES C1CC1C(=O)C2=CC=C(C=C2)Br

Spécifications

Melting Point 43°C
Color White-Yellow
Boiling Point 204°C
Quantity 5 g
Formula Weight 225.09
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Bromophenyl Cyclopropyl Ketone