missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Bromomethyl)-3-fluorobenzonitrile 98.0+%, TCI America™
Supplier: TCI America B320425G
Specifications
| 4-(Bromomethyl)-3-fluorobenzonitrile | |
| 77°C | |
| C8H5BrFN | |
| 25 g | |
| 4-bromomethyl-3-fluorobenzonitrile, 4-cyano-2-fluorobenzyl bromide, 2-fluoro-4-cyanobenzyl bromide, 2-fluoro-4-cyanbenzyl bromide, benzonitrile, 4-bromomethyl-3-fluoro, 2-fluoro-4-cyanobenzylbromide, 3-fluoro-4-bromomethyl benzonitrile, 4-bromomethyl-3-fluoro-benzonitrile, 4-bromomethyl-3-fluorobenzenecarbonitrile, pubchem4907 | |
| FC1=C(CBr)C=CC(=C1)C#N | |
| 214.04 | |
| 214.04 | |
| Crystalline Powder |
| 105942-09-4 | |
| White-Yellow | |
| MFCD07368341 | |
| 3439 | |
| ZESZAIOGACKOMB-UHFFFAOYSA-N | |
| 4-(bromomethyl)-3-fluorobenzonitrile | |
| 2783149 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 105942-09-4 | |
| 214.04 | |
| ZESZAIOGACKOMB-UHFFFAOYSA-N | |
| 2783149 | |
| FC1=C(CBr)C=CC(=C1)C#N |
| C8H5BrFN | |
| MFCD07368341 | |
| 4-bromomethyl-3-fluorobenzonitrile, 4-cyano-2-fluorobenzyl bromide, 2-fluoro-4-cyanobenzyl bromide, 2-fluoro-4-cyanbenzyl bromide, benzonitrile, 4-bromomethyl-3-fluoro, 2-fluoro-4-cyanobenzylbromide, 3-fluoro-4-bromomethyl benzonitrile, 4-bromomethyl-3-fluoro-benzonitrile, 4-bromomethyl-3-fluorobenzenecarbonitrile, pubchem4907 | |
| 4-(bromomethyl)-3-fluorobenzonitrile |
Safety and Handling
TSCA : No