accessibility menu, dialog, popup

UserName

CAS RN 105942-09-4

IUPAC Name: 4-(bromomethyl)-3-fluorobenzonitrile
Synonyms: 4-(bromomethyl)-3-fluorobenzonitrile
Molecular Weight (g/mol): 214.04
Molecular Formula: C8H5BrFN
InChi Key: ZESZAIOGACKOMB-UHFFFAOYSA-N
SMILES: FC1=C(CBr)C=CC(=C1)C#N
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)