missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)-3-fluorobenzonitrile 98.0+%, TCI America™

Catalog No. p-7122806
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32041G 1 g
B32045G 5 g
B320425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B32041G Supplier TCI America Supplier No. B32041G
Only null left

Chemical Identifiers

CAS 105942-09-4
Molecular Formula C8H5BrFN
Molecular Weight (g/mol) 214.04
MDL Number MFCD07368341
InChI Key ZESZAIOGACKOMB-UHFFFAOYSA-N
Synonym 4-bromomethyl-3-fluorobenzonitrile, 4-cyano-2-fluorobenzyl bromide, 2-fluoro-4-cyanobenzyl bromide, 2-fluoro-4-cyanbenzyl bromide, benzonitrile, 4-bromomethyl-3-fluoro, 2-fluoro-4-cyanobenzylbromide, 3-fluoro-4-bromomethyl benzonitrile, 4-bromomethyl-3-fluoro-benzonitrile, 4-bromomethyl-3-fluorobenzenecarbonitrile, pubchem4907
PubChem CID 2783149
IUPAC Name 4-(bromomethyl)-3-fluorobenzonitrile
SMILES FC1=C(CBr)C=CC(=C1)C#N

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 214.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Bromomethyl)-3-fluorobenzonitrile